Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gp210 Rabbit anti-Mammalia, Rat, Xenopus laevis, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA15781

Catalog No. 01-674-972


Add to cart

Description

Description

PA1-5781 detects gp210, NT in mammalian, rat and xenopus samples. PA1-5781 has been successfully used in Western blot procedures. The PA1-5781 immunogen is purified bacterial N-terminal fusion protein.

The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.
Specifications

Specifications

gp210
Polyclonal
whole serum with 0.1% sodium azide
Q8TEM1, Q9QY81, P11654
FLJ22389, GP210, KIAA0906, POM210
Rabbit
IgG
RUO
Antibody
Polyclonal
Mammalia, Rat, Xenopus laevis
Western Blot
Unconjugated
NUP210
Liquid
NUP210, nup210.L
Purified bacterial N-terminal fusion protein.
100 μL
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
23225, 398395, 54563, 58958
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit