Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

gp210 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA15782

Catalog No. 01-674-973



Description

Description

PA1-5782 detects gp210, CT in rat samples, and cross-reacts with mouse, human, and monkey. PA1-5782 has been successfully used in Western blot and immunofluorescence procedures. The PA1-5782 immunogen is purified bacterial C-terminal fusion protein.

The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.
Specifications

Specifications

gp210
Polyclonal
whole serum with 0.02% sodium azide
Q8TEM1, Q9QY81, P11654
FLJ22389, GP210, KIAA0906, POM210
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
NUP210
Liquid
NUP210
Purified bacterial C-terminal fusion protein
100 μL
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
23225, 54563, 58958
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit