Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

glycoprotein IX (platelet), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human GP9 protein.

Manufacturer:  Abnova Corporation H00002815B01

Catalog No. 89-005-462


Add to cart

Description

Description

This gene encodes a small membrane glycoprotein found on the surface of human platelets. It forms a 1-to-1 noncovalent complex with glycoprotein Ib, a platelet surface membrane glycoprotein complex that functions as a receptor for von Willebrand factor. The complete receptor complex includes noncovalent association of the alpha and beta subunits with the protein encoded by this gene and platelet glycoprotein V. Defects in this gene are a cause of Bernard-Soulier syndrome, also known as giant platelet disease. These patients have unusually large platelets and have a clinical bleeding tendency. [provided by RefSeq

Sequence: MPAWGALFLLWATAEATKDCPSPCTCRALETMGLWVDCRGHGLTALPALPARTRHLLLANNSLQSVPPGAFDHLPQLQTLDVTQNPWHCDCSLTYLRLWLEDRTPEALLQVRCASPSLAAHGPLGRLTGYQLGSCGWQLQASWVRPGVLWDVALVTVAALGLALLAGLLCATTEALD
Specifications

Specifications

glycoprotein IX (platelet)
Unconjugated
No additive
BC030229.1
CD42a|GPIX
Mouse
50 uL
Cell Adhesion
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human GP9 protein.
GP9
Serum
GP9
GP9 (AAH30229.1, 1 a.a. ~ 177 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
2815
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit