Learn More
GPAA1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524215
Catalog No. PIPA524215
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Posttranslational glycosylphosphatidylinositol (GPI) anchor attachment serves as a general mechanism for linking proteins to the cell surface membrane. The protein encoded by this gene presumably functions in GPI anchoring at the GPI transfer step. The mRNA transcript is ubiquitously expressed in both fetal and adult tissues. The anchor attachment protein 1 contains an N-terminal signal sequence, 1 cAMP- and cGMP-dependent protein kinase phosphorylation site, 1 leucine zipper pattern, 2 potential N-glycosylation sites, and 8 putative transmembrane domains.
Specifications
Specifications
GPAA1
Polyclonal
Unconjugated
GPAA1
Liquid
GPAA1
KLH conjugated synthetic peptide between 46-73 amino acids from the N-terminal region of human GPAA1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.26 mg/mL
PBS with 0.09% sodium azide
O43292
GAA1, hGAA1, GAA1 protein homolog, GPAA1P anchor attachment protein 1 homolog, GPI anchor attachment protein 1, GPI transamidase subunit, anchor attachment protein 1 (Gaa1p, yeast) homolog, glycophosphatidylinositol anchor attachment 1, glycosylphosphatidylinositol anchor a
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8733