Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GPAM, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232116

Catalog No. NBP232116


Add to cart

Description

Description

GPAM Polyclonal antibody specifically detects GPAM in Human samples. It is validated for Western Blot.
Specifications

Specifications

GPAM
1.0 mg/mL
Western Blot 1:500 - 1:2500
NP_001231878.1
EC 2.3.1, EC 2.3.1.15, glycerol-3-phosphate acyltransferase 1, mitochondrial, glycerol-3-phosphate acyltransferase, mitochondrial, GPAT, GPAT-1, GPAT1mitochondrial, MGC26846
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
GPAM
The immunogen this antibody was made to, maps to a region between residue 275 to 325 of human Glycerol-3-phosphate Acyltransferase, Mitochondrial using the numbering given in entry NP_001231878.1 (GeneID 57678).
Immunogen affinity purified
RUO
Primary
57678
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only