Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPAM Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523659

Catalog No. PIPA523659

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Glycerol-3-phosphate acyltransferase (GPAT; EC 2.3.1.15),which catalyzes the initial and committing step in glycerolipidbiosynthesis, is predicted to play a pivotal role in the regulationof cellular triacylglycerol and phospholipid levels. Two mammalianforms of GPAT have been identified on the basis of localization toeither the endoplasmic reticulum or mitochondria.[supplied byOMIM].
Specifications

Specifications

GPAM
Polyclonal
PBS with 0.09% sodium azide
Q9HCL2
GPAT, GPAT1, GPAT-1, glycerol 3-phosphate acyltransferase, mitochondrial, glycerol-3-phosphate acyltransferase 1, mitochondrial
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57678
Flow Cytometry, Western Blot
Unconjugated
GPAM
Liquid
GPAM
KLH conjugated synthetic peptide between 426-455 amino acids from the central region of human GPAM
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit