Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPATCH1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP191957

Catalog No. NBP191957


Add to Cart

Description

Description

GPATCH1 Polyclonal specifically detects GPATCH1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

GPATCH1
Polyclonal
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
ECGPFLJ34516, evolutionarily conserved G patch domain containing, evolutionarily conserved G-patch domain containing, Evolutionarily conserved G-patch domain-containing protein, FLJ10206, FLJ38686, G patch domain containing 1, G patch domain-containing protein 1, GPATC1
Rabbit
Affinity Purified
RUO
55094
Human
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
GPATCH1
This antibody was developed against Recombinant Protein corresponding to amino acids:DVTPVDFTPKDNVHGLAYKGLDPHQALFGTSGEHFNLFSGGSERAGDLGEIGLNKGRKLGISGQAFGVGALEEEDDDIYATETLSKYDTVLKDE
0.1 mL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only