Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPATCH4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179018
Description
GPATCH4 Polyclonal specifically detects GPATCH4 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
GPATCH4 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
G patch domain containing 4 | |
Rabbit | |
Affinity Purified | |
RUO | |
54865 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation | |
EAW52937.1 | |
GPATCH4 | |
The immunogen recognized by this antibody maps to a region between residue 396 and 446 of human G Patch Domain Containing 4 using the numbering given in entry EAW52937.1 (GeneID 54865). | |
0.1 mL | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction