Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPI-Phospholipase D Mouse anti-Human, Unlabeled, Clone: 15, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610810

Catalog No. BDB610810


Add to cart

Description

Description

Glycosylphosphatidylinositol (GPI)-anchored proteins are involved in the regulation of numerous cell-surface activities including the complement cascade, activation of plasminogen, T-cell activation, cell-to-cell adhesion, and macromolecule transport through the membrane. GPI-containing proteins are very abundant at the caveolae. GPI-specific phospholipase D (GPI-PLD) hydrolyzes the covalently bound GPI moiety which functions as an anchor for many cell-surface proteins. Present at high levels in the plasma, GPI-PLD is also secreted by some cell types in culture. This protein releases the GPI-anchored, complement regulatory protein decay accelerating factor (DAF) from HeLa cells, as well as the basic fibroblast growth factor (bFGF)-bound heparin sulfate proteoglycan from bone marrow stromal cells. GPI-PLD contains Ca2+; and Zn2+; binding sites and bears some further structural and perhaps functional similarities to the integrins.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

GPI-Phospholipase D
15
Unconjugated
Affinity Purified
Human GPI-Phospholipase D aa.706-841
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit