Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIGK Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512177

Catalog No. PIPA512177

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

GPI8 is a member of the cysteine protease family C13 that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is a member of the multisubunit enzyme, GPI transamidase and is thought to be its enzymatic component. GPI transamidase mediates GPI anchoring in the endoplasmic reticulum, by catalyzing the transfer of fully assembled GPI units to proteins.
Specifications

Specifications

PIGK
Polyclonal
PBS with 0.09% sodium azide
Q92643, Q9CXY9
GPI-anchor transamidase; GPI transamidase; Phosphatidylinositol-glycan biosynthesis, class K protein; PIG-K; hGPI8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10026, 329777
Immunohistochemistry, Western Blot
Unconjugated
PIGK
Liquid
PIGK
KLH conjugated synthetic peptide between 266-297 amino acids from the C-terminal region of human GPI8
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit