Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPIHBP1 Rabbit anti-Bovine, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA116976

Catalog No. 01-670-153


Add to cart

Description

Description

Suggested positive control: mouse kidney lysate or protein.

GPI-anchored HDL-binding protein 1 (Gpihbp1) is a novel protein that may be involved in the regulation of the delivery of fats to cells for energy and storage. Digested fats travel to the small intestine, where they are packaged into chylomicrons (particles filled with triglycerides). Chylomicrons then travel through the bloodstream and deliver triglycerides to tissues that are hungry for fuel or to adipose tissue for energy storage. Triglycerides are broken down or hydrolyzed by the enzyme lipoprotein lipase (LpL). The triglyceride breakdown products are then taken up and used by cells. Gpihbp1 is the molecule in capillaries that facilitates the capture of chylomicrons and facilitates the interaction with LpL. It has been shown that fats in the bloodstream are not readily metabolized in the absence of GPIHBP1.
Specifications

Specifications

Gpihbp1
Polyclonal
Unconjugated
Gpihbp1
Liquid
GPIHBP1
Synthetic peptide corresponding to residues 100-170 of mouse GPI-anchored HDL-binding protein 1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.6 mg/mL
tris glycine with 0.15M NaCl and 0.05% sodium azide
Q9D1N2
GPI-anchored HDL-binding protein 1 antibody, GPI-anchored High-Density Lipid-binding protein 1 antibody, GlycosylPhosphatidylInositol-anchored high-de
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
300027, 512826, 68453
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit