Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

glycosylphosphatidylinositol specific phospholipase D1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human GPLD1 protein.

Manufacturer:  Abnova Corporation H00002822B01

Catalog No. 89-005-465


Add to cart

Description

Description

Many proteins are tethered to the extracellular face of eukaryotic plasma membranes by a glycosylphosphatidylinositol (GPI) anchor. The GPI-anchor is a glycolipid found on many blood cells. The protein encoded by this gene is a GPI degrading enzyme. Glycosylphosphatidylinositol specific phospholipase D1 hydrolyzes the inositol phosphate linkage in proteins anchored by phosphatidylinositol glycans, thereby releasing the attached protein from the plasma membrane. [provided by RefSeq

Sequence: MSAFRLWPGLLIMLGSLCHRGSPCGLSTHIEIGHRALEFLQLHNGRVNYRELLLEHQDAYQAGIVFPDCFYPSICKGGKFHDVSESTHWTPFLNASVHYIRENYPLPWEKDTEKLVAFLFGITSHMAADVSWHSLGLEQGFLRTMGAIDFHGSYSEAHSAGDFGTVYLHLLNFLVV
Specifications

Specifications

glycosylphosphatidylinositol specific phospholipase D1
Unconjugated
No additive
BC020748.1
GPIPLD|GPIPLDM|MGC22590|PIGPLD|PIGPLD1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
2822
Western Blot
Mouse polyclonal antibody raised against a full-length human GPLD1 protein.
GPLD1
Serum
GPLD1
GPLD1 (AAH20748.1, 1 a.a. ~ 176 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit