Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPM6A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526350

Catalog No. PIPA526350

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

Specifications

Specifications

GPM6A
Polyclonal
PBS with 0.09% sodium azide
P51674
GPM6, M6A, neuronal membrane glycoprotein M6-a
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2823
Flow Cytometry, Western Blot
Unconjugated
GPM6A
Liquid
GPM6A
KLH conjugated synthetic peptide between 232-259 amino acids from the C-terminal region of human GPM6A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit