Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GPR10, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP216716

Catalog No. NBP216716


Add to cart

Description

Description

GPR10 Polyclonal specifically detects GPR10 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

GPR10
1mg/mL
Western Blot 1:500-1:3000
Affinity Purified
PRLHR
Recombinant protein encompassing a sequence within the center region of human GPR10. The exact sequence is proprietary.
41 kDa
0.1 ml
GPCR
Primary
2834
Western Blot
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
G protein-coupled receptor 10, GPR10MGC126539, G-protein coupled receptor 10, GR3, hGR3, MGC126541, prolactin releasing hormone receptor, prolactin releasing peptide receptor, prolactin-releasing hormone receptor, prolactin-releasing peptide receptor, PrRP receptor, PrRPR
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only