Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR133 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526508

Catalog No. PIPA526508

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GPR133 is an orphan receptor.
Specifications

Specifications

GPR133
Polyclonal
PBS with 0.09% sodium azide
Q6QNK2
PGR25, G-protein coupled receptor PGR25, probable G-protein coupled receptor 133
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
283383
Western Blot
Unconjugated
ADGRD1
Liquid
ADGRD1
KLH conjugated synthetic peptide between 471-498 amino acids from the central region of human GPR133
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit