Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GPR155, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP150626H

Catalog No. NBP150626H


Add to cart

Description

Description

GPR155 Polyclonal antibody specifically detects GPR155 in Human samples. It is validated for Western Blot.
Specifications

Specifications

GPR155
HRP
PBS with No Preservative
Affinity Purified
GPR155
A synthetic peptide made to an internal portion of the human GPR155 protein (between residues 800-900) [UniProt Q7Z3F1]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human
Western Blot
Western Blot
Q7Z3F1
DEP.7, DEPDC3, FLJ31819, FLJ39346, G protein-coupled receptor 155, integral membrane protein GPR155, PGR22G-protein coupled receptor PGR22
Rabbit
IgG
0.1 ml
Cancer, Neuroscience, Signal Transduction, Stem Cell Lines
Primary
151556
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only