Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR155 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523074

Catalog No. PIPA523074


Add to cart

Description

Description

The target sequence has 86% sequence homology with mouse.

GPR155, an integral membrane protein related to G-protein coupled receptors, has been shown to have specific roles in Huntington disease and autism spectrum disorders.
Specifications

Specifications

GPR155
Polyclonal
Unconjugated
GPR155
Liquid
GPR155
A synthetic peptide made to an internal portion of the human GPR155 protein (between residues 500-600)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1.06 mg/mL
PBS with 30% glycerol and 0.05% sodium azide
Q7Z3F1
DEP.7, DEPDC3, PGR22, G-protein coupled receptor PGR22, integral membrane protein GPR155
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
151556
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit