Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR17 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526851

Catalog No. PIPA526851

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Members of the G protein coupled receptor (GPCR) superfamily contain 7 trans membrane domains and transduce extracellular signals through heterotrimeric G proteins. The organization of the GPR17 gene differs from that of many other GPCRs in that the open reading frame is distributed on 2 exons; an additional exon contains the 5 prime untranslated region. Human GPR17 is expressed as 2.3 and 6.3 kb mRNAs exclusively in brain. The 2 transcripts appear to represent alternatively polyadenylated variants. Based on protein sequence homology and the conservation of certain key residues, GPR17 appears to be closely related to the P2Y family of GPCRs. There are two isoforms.
Specifications

Specifications

GPR17
Polyclonal
PBS with 0.09% sodium azide
Q13304
G-protein coupled receptor 17, P2Y-like receptor, R12, UDP/CysLT receptor, uracil nucleotide/cysteinyl leukotriene receptor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2840
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
GPR17
Liquid
GPR17
KLH conjugated synthetic peptide between 230-258 amino acids from the central region of human GPR17
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit