Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR180 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526788
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
This gene encodes a protein that is a member of the G protein-coupled receptor superfamily. This protein is produced predomitly in vascular smooth muscle cells and may play an important role in the regulation of vascular remodeling.Specifications
GPR180 | |
Polyclonal | |
Unconjugated | |
Q86V85 | |
GPR180 | |
KLH conjugated synthetic peptide between 119-147 amino acids from the central region of human GPR180 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
GPR180 | |
ITR, integral membrane protein GPR180, intimal thickness-related receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
160897 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction