Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR50 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526352

Catalog No. PIPA526352

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Melatonin-related receptor expression has been documented from human CNS tissues, specifically in hypothalamus and pituitary, but not in human peripheral tissues. In animals, expression has also been seen in the CNS, and peripheral tissues.
Specifications

Specifications

GPR50
Polyclonal
PBS with 0.09% sodium azide
Q13585
H9, Mel1c, melatonin-related receptor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9248
Flow Cytometry, Western Blot
Unconjugated
GPR50
Liquid
GPR50
KLH conjugated synthetic peptide between 316-344 amino acids from the central region of human GPR50
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit