Learn More
anti-GPR68, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232747
Catalog No. NBP232747
Add to cart
Description
Description
OGR1 Polyclonal antibody specifically detects OGR1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
GPR68
0.5 mg/mL
Western Blot 1:1000
Q8BFQ3
G protein-coupled receptor 68, GPR12A, G-protein coupled receptor 68, MGC111379, OGR-1, OGR1MGC156983, ovarian cancer G protein-coupled receptor, 1, ovarian cancer G-protein coupled receptor 1, Sphingosylphosphorylcholine receptor
Rabbit
IgG
Protein A purified
RUO
Primary
8111
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
GPR68
This Mouse GPR68 Antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 200-225 amino acids from the Central region of mouse Gpr68.
41.209 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse