Learn More
anti-GPR83, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP133001
Catalog No. NBP133001
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
GPR83
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
GPR83
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GPR83. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific to the human GPR72(GPR83) protein.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
G protein-coupled receptor 72, G protein-coupled receptor 83, GIR, GPR72glucocorticoid induced receptor, G-protein coupled receptor 72, KIAA1540, probable G-protein coupled receptor 83
Rabbit
IgG
100 ul
GPCR
Primary
10888
Human