Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR83 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527259

Catalog No. PIPA527259


Add to cart

Description

Description

PA5-27259 targets GPR83 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27259 immunogen is synthetic peptide corresponding to a region within amino acids 357 and 423 of Human GPR83. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Orphan receptor. Could be a neuropeptide Y receptor.
Specifications

Specifications

GPR83
Polyclonal
Unconjugated
GPR83
Liquid
GPR83
Synthetic peptide corresponding to a region within amino acids 357 and 423 of Human GPR83
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9NYM4
GIR, GPR72, G protein-coupled receptor 72, G-protein coupled receptor 72, glucocorticoid induced receptor, probable G-protein coupled receptor 83
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10888
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit