Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPSN2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526789

Catalog No. PIPA526789

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2,3-enoyl-CoA to saturated acyl-CoA.
Specifications

Specifications

GPSN2
Polyclonal
PBS with 0.09% sodium azide
Q9NZ01
GPSN2, MRT14, SC2, TER, glycoprotein, synaptic 2, synaptic glycoprotein SC2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9524
Immunohistochemistry, Western Blot
Unconjugated
TECR
Liquid
TECR
KLH conjugated synthetic peptide between 268-295 amino acids from the C-terminal region of human GPSN2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit