Learn More
Platelet GPVI Mouse anti-Human, Alexa Fluor 647, Clone: HY101 (also known as HY 101; HY-101), BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564701
Catalog No. BDB564701
Add to cart
Description
Description
The HY101 monoclonal antibody specifically binds to a collagen receptor, Platelet Glycoprotein VI (GPVI or GP6). GPVI is a 58-kD platelet membrane glycoprotein that belongs to the immunoglobulin superfamily. Upon blood vessel wall injury and consequent endothelial cell damage, platelets collect upon the exposed collagen fibers present in the subendothelial matrix. The platelet and collagen interaction leads to activated platelet aggregation and thrombus formation. Although normally serving a protective function in clot formation that limits blood loss, the platelet GPVI/collagen interaction may also play a role in myocardial infarction and stroke which are associated with the rupture of atherosclerotic plaques.
Specifications
Specifications
Platelet GPVI
HY101 (also known as HY 101; HY-101)
Glycoprotein 6; Platelet glycoprotein VI; Platelet collagen receptor
Affinity Purified
Glycoprotein VI Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal