Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-GRAMD1A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232146

Catalog No. NBP232146



Description

Description

GRAMD1A Polyclonal antibody specifically detects GRAMD1A in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

GRAMD1A
1.0 mg/mL
Immunoprecipitation 2 - 10 ug/mg lysate
NP_065946.2
FLJ22411, FLJ90346, GRAM domain containing 1A, GRAM domain-containing protein 1A, KIAA1533
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
GRAMD1A
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human GRAM Domain Containing 1A using the numbering given in entry NP_065946.2 (GeneID 57655).
Immunogen affinity purified
RUO
Primary
57655
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only