Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRAM domain containing 1B, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant KIAA1201.

Manufacturer:  Abnova Corporation H00057476A01

Catalog No. 89-004-696


Add to cart

Description

Description

Sequence: NTMGYCEEIPVEENEVNDSSSKSSIETKPDASPQLPKKSITNSTLTSTGSSEAPVSFDGLPLEEEALEGDGSLEKELAIDNIMGEKIEMIAPVNSPSLDFNDNE
Specifications

Specifications

GRAM domain containing 1B
Unconjugated
50% glycerol
XM_370660
KIAA1201|MGC125622|MGC125623|MGC125625
Mouse
50 uL
Signal Transduction
Y
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant KIAA1201.
GRAMD1B
Serum
GRAMD1B
KIAA1201 (XP_370660, 250 a.a. ~ 353 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
57476
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit