Learn More
anti-Granulin, Polyclonal, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 263200020.1MG
 View more versions of this product
Catalog No. 26-320-00201
  Check Availability
Add to cart
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
Granulin
Unlabeled
P28799
Rabbit
0.1mg
Primary
2896
Human,Primate
Blocking,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Neutralization,Western Blot
20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
Affinity Purified
This antibody is specific for the Middle Region of the target protein.
RUO
Polyclonal
This product is specific for Human grn. Human. primate reactivity reported in scientific literature (PMID: 24163244).