Learn More
GM-CSF, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA184988
Catalog No. PIPA184988
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
GM-CSF (granulocyte-macrophage colony-stimulating factor) is a 14.6kDa haematopoietic growth factor which exists in both glycosylated and non-glycosylated biologically active forms, and stimulates the development of granulocytes, macrophages, early megakaryocytes and eosinophil progenitor cells. The ability of recombinant GM-CSF to increase haematopoietic cell recovery has become a focus area in the therapeutic treatment of patients following bone marrow transplantation.
Reconstitute with 0.5 ml PBS containing 0.1% BSA. For long term storage the addition of 0.09% sodium azide is recommended. PA1-84988 has a IgG concentration of ~0.1 mg/ml after reconstitution. Prior to reconstitution, store at 4ºC. After reconstitution, store undiluted at -20ºC, avoiding freeze/thaw cycles.This antibody may be used in a direct ELISA or as the detection reagent in a sandwich ELISA with PA1-84972 as the capture antibody.
Specifications
Specifications
GM-CSF
Polyclonal
Biotin
CSF2
Lyophilized
CSF2
Recombinant protein GM-CSF.
Affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
0.1mg/mL
PBS with no preservative
P04141
Csfgm, Gm-CSf, MGC151255, MGC151257, MGI-IGM
Rabbit
IgG
50μg
4° C
Primary
1437