Learn More
GM-CSF, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA184972
Catalog No. PA184972
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
GM-CSF (granulocyte-macrophage colony-stimulating factor) is a 14.6kDa haematopoietic growth factor which exists in both glycosylated and non-glycosylated biologically active forms, and stimulates the development of granulocytes, macrophages, early megakaryocytes and eosinophil progenitor cells. The ability of recombinant GM-CSF to increase haematopoietic cell recovery has become a focus area in the therapeutic treatment of patients following bone marrow transplantation.
Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections.Reconstitute with 0.1 ml of distilled water. For long term storage 0.09% sodium azide is recommended. After reconstitution, store at -20ºC.This antibody may be used in an indirect ELISA or as a capture reagent in a sandwich ELISA with PA1-84988 as the detection antibody.
Specifications
Specifications
GM-CSF
Polyclonal
Unconjugated
CSF2
Lyophilized
CSF2
Recombinant human GM-CSF
Affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Functional Assay, Immunohistochemistry (Paraffin), Western Blot
1mg/mL
PBS with no preservative; pH 7.4
P04141
GMCSF, MGC131935, MGC138897
Rabbit
IgG
100μg
4° C
Primary
1437