Learn More
GRB2-related adaptor protein, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human GRAP protein.
Manufacturer: Abnova Corporation H00010750B01
Description
This gene encodes a member of the GRB2/Sem5/Drk family. This member functions as a cytoplasmic signaling protein which contains an SH2 domain flanked by two SH3 domains. The SH2 domain interacts with ligand-activated receptors for stem cell factor and erythropoietin, and facilitates the formation of a stable complex with the BCR-ABL oncoprotein. This protein also associates with the Ras guanine nucleotide exchange factor SOS1 (son of sevenless homolog 1) through its N-terminal SH3 domain. [provided by RefSeq
Sequence: MESVALYSFQATESDELAFNKGDTLKILNMEDDQNWYKAELRGVEGFIPKNYIRVKPHPWYSGRISRQLAEEILMKRNHLGAFLIRESESSPGEFSVSVNYGDQVQHFKVLREASGKYFLWEEKFNSLNELVDFYRTTTIAKKRQIFLRDEEPLLKSPGACFAQAQFDFSAQDPSQLSFRRGDIIEVLERPDPHWWRGRSCGRVGFFPRSYVQPVHLSpecifications
GRB2-related adaptor protein | |
Unconjugated | |
No additive | |
NM_006613 | |
MGC64880 | |
Mouse | |
50 uL | |
Membrane Receptors | |
Y | |
Polyclonal | |
Human |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human GRAP protein. | |
GRAP | |
Serum | |
GRAP | |
GRAP (NP_006604, 1 a.a. ~ 217 a.a) full-length human protein. | |
RUO | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
10750 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.