Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRASP55 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1076

Catalog No. 01-670-190


Add to cart

Description

Description

PA1-076 detects GRASP55 from mouse and rat samples. PA1-076 has been successfully used in Western blot applications. By Western blot this antibody detects a ~55 kDa band representing the GRASP55 protein. The PA1-076 immunogen is a synthetic peptide corresponding to residues P(419) A S K V P T T V E D R V S D(433) of rat GRASP55 protein. This peptide is available for neutralization controls as PEP-295.

GRASP55 is a PDZ containing protein involved in the stacking of golgi cisternea in a cell-free system.
Specifications

Specifications

GRASP55
Polyclonal
Unconjugated
GORASP2
Liquid
GORASP2
Synthetic peptide corresponding to residues P(419) A S K V P T T V E D R V S D(433) of rat GRASP55 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse, Rat
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q9H8Y8, Q99JX3, Q9R064
Golgi reassembly-stacking protein 2, GRS2, Golgi reassembly-stacking protein of 55 kDa
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
113961, 26003, 70231
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit