Learn More
GRB2 Mouse anti-Bovine, Human, Hamster, Mouse, Rat, Clone: 3F2, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05372
Catalog No. 05-372-MI
Add to cart
Description
Description
This Anti-GRB2 Antibody, clone 3F2 is validated for use in immunohistochemistry, western blotting for the detection of GRB2.
Specifications
Specifications
GRB-2
Monoclonal
Unconjugated
NM_002086
Mouse
Protein A chromatography
Primary
−20°C for 24 months.
IgG1 κ
Immunohistochemistry, Western Blot
3F2
PBS containing 50% glycerol
GRB2, EGFRBP-GRB2, HT027, ASH, MST084, MSTP084, Grb3-3
Recombinant GRB2/Ash
100 μg
Bovine, Hamster, Human, Mouse, Rat
Purified