Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat GRB2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3846
This item is not returnable.
View return policy
Description
GRB2 Monoclonal specifically detects GRB2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GRB2 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P62993 | |
GRB2 | |
E. coli-derived recombinant human GRB2 Met1-Val217 Accession # P62993 | |
100 μg | |
Primary | |
Detects human, mouse and rat GRB2 in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
481021 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
abundant SRC homology, Adapter protein GRB2, ASH, EGFRBP-GRB2, epidermal growth factor receptor-binding protein GRB2, Grb3-3, growth factor receptor-bound protein 2, growth factor receptor-bound protein 3, HT027, MST084, MSTP084, NCKAP2, Protein Ash, SH2/SH3 adapter GRB2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2885 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction