Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRB2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513280

Catalog No. PIPA513280

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken, mouse, rat and Xenopus based on sequence homology.

GRB2 binds the epidermal growth factor receptor and contains one SH2 domain and two SH3 domains. Its two SH3 domains direct complex formation with proline-rich regions of other proteins, and its SH2 domain binds tyrosine phosphorylated sequences. This gene is similar to the Sem5 gene of C.elegans, which is involved in the signal transduction pathway.
Specifications

Specifications

GRB2
Polyclonal
PBS with 0.09% sodium azide
P62993
Growth factor receptor-bound protein 2; Adapter protein GRB2; SH2/SH3 adapter GRB2; Protein Ash; ASH
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2885
Immunohistochemistry, Western Blot
Unconjugated
GRB2
Liquid
GRB2
KLH conjugated synthetic peptide between 187-216 amino acids from human GRB2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit