Learn More
MilliporeSigma™ anti-Green Fluorescent Protein (26-39), Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ PC408100UL
Catalog No. PC408100UL
Add to cart
Description
Description
Specifically detects Green Fluorescent Protein (26-39) Clone: in samples, and it is validated for Immunocytochemistry
Green Fluorescent Protein (GFP) is responsible for bioluminescence in the jellyfishAequorea victoria. It has quickly become a powerful research tool for assessing gene expression and subcellular protein distribution in fixed or living cells. GFP is excited by and brightly fluoresces when exposed to UV or blue light. This feature makes it ideal as a marker for use in fluorescence microscopy, cytometry, tagging fusion proteins, and assaying transcriptional regulation from regulation from gene promotersin vivo. Numerous GFP variants with enhanced and shifted emission spectra (blue, green, and yellow) have been developed through amino acid substitutions at specific residues.
Specifications
Specifications
Green Fluorescent Protein (26-39)
Unconjugated
purified
a synthetic peptide corresponding to amino acids 26-39 of Aequorea victoria wild-type green fluorescent protein
100μL
−20°C
Polyclonal
Immunocytochemistry
In PBS, 1mg/mL BSA.
Rabbit
IgG
RUO
Primary
Recognizes the ∽28kDa GFP and some GFP fusion proteins.