Learn More
MilliporeSigma™ anti-Green Fluorescent Protein (GFP); Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB16901/DEL
Description
The Green Fluorescent Protein (GFP) from the jellyfish Aequorea victoria is used as a fluorescent indicator for monitoring gene expression in a variety of cellular systems, including living organisms and fixed tissues. Unlike other bioluminescent reporters, GFP fluoresces in the absence of substrates, cofactors, or other intrinsic or extrinsic proteins. Purified GFP is a 27 kDa monomer consisting of 238 amino acids and emits green light (emission maximum at 509nm) when excited with blue or UV light.
Immunocytochemistry, Immunoprecipitation, Western Blotting
Specifications
GFP (Green Fluorescent Protein) | |
Unconjugated | |
A recombinant green fluorescent protein (GFP) containing a 6-his tag, was highly purified by affinity chromatography on Nickel column followed by chromatography on a P-100 gel filtration column. | |
RUO | |
Primary | |
Semi-purified |
Polyclonal | |
Chicken | |
250 μg | |
Epitope Tags | |
Vertebrates |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.