Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520453

Catalog No. PIPA520453


Add to cart

Description

Description

Despite its predicted molecular weight, Grik4 often migrates at a lower molecular weight in SDS-PAGE. A suggested positive control is rat brain tissue lysate. PA5-20453 can be used with blocking peptide PEP-0570.

Grik4 codes for the KA1 subunit of kainate-type ionotropic glutamate receptors which are critical regulators of network activity that act by modifying neuronal excitability, directly and indirectly, through GABAergic interneurons. Five subunits can assemble to form kainate receptors (KARs): GluR5 (coded by Grik1), GluR6 (coded by Grik2), and GluR7 (coded by Grik3) are the low-affinity subunits, and KA1 and KA2 are the high-affinity subunits. In the adult brain, KARs are located pre- and postsynaptically on pyramidal cells and on interneurons. Kainate receptors on GABA-containing interneurons enhance GABA release and thereby downregulate glutamatergic signaling. KARs have been implicated in numerous psychiatric disorders. Case control studies show significant association of Grik4 with both schizophrenia and bipolar disorder.
Specifications

Specifications

Grik4
Polyclonal
Unconjugated
GRIK4
Liquid
GRIK4
A 14 amino acid peptide near the amino terminus of the human Grik4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q16099, Q8BMF5, Q01812
EAA1, GRIK, KA1; excitatory amino acid receptor 1; glutamate receptor KA-1; glutamate receptor KA1; glutamate receptor, ionotropic kainate 4
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
110637, 24406, 2900
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit