Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIN2C, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human GRIN2C protein.

Manufacturer:  Abnova Corporation H00002905D01

Catalog No. 89-002-892


Add to cart

Description

Description

N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C), and NMDAR2D (GRIN2D).

Sequence: MGGALGPALLLTSLFGAWAGLGPGQGEQGMTVAVVFSSSGPPQAQFRARLTPQSFLDLPLEIQPLTVGVNTTNPSSLLTQICGLLGAAHVHGIVFEDNVDTEAVAQILDFISSQTHVPILSISGGSAVVLTPKVHVQTHVPSCLRPGTRLGSGVLWFWEAGIRRDGQGGGG
Specifications

Specifications

GRIN2C
Unconjugated
No additive
BC059384
NMDAR2C|NR2C
Rabbit
100 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunoprecipitation, Western Blot
Rabbit polyclonal antibody raised against a full-length human GRIN2C protein.
GRIN2C
Liquid
GRIN2C
GRIN2C (AAH59384.1, 1 a.a. ~ 171 a.a) full-length human protein.
RUO
Primary
2905
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit