Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRINA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523994

Catalog No. PIPA523994


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken, mouse, porcine, non-human primate, rat, rabbit and zebrafish based on sequence homology.

Specifications

Specifications

GRINA
Polyclonal
PBS with 0.09% sodium azide
Q7Z429
HNRGW, LFG1, NMDARA1, TMBIM3, NMDA receptor glutamate-binding subunit, glutamate [NMDA] receptor-associated protein 1, glutamate receptor, NMDA subtype, glutamate-binding subunit, putative MAPK-activating protein PM02, transmembrane BAX inhibitor motif containing 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2907
Western Blot
Unconjugated
GRINA
Liquid
GRINA
KLH conjugated synthetic peptide between 117-146 amino acids from the central region of human GRINA
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit