Learn More
MilliporeSigma™ anti-GRK6 Clone: 2C3, Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ ST1651100UG
Catalog No. ST1651100UG
Add to cart
Description
Description
Specifically detects GRK6 Clone: 2C3 in Human samples, and it is validated for ELISA, Immuno Blotting
This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. Several transcript variants encoding different isoforms have been described for this gene.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
GRK6
2C3
In PBS, pH 7.2.
Murine
IgG2a
RUO
Primary
Recognizes the ∽65-70kDa GRK6 protein in HeLa and Jurkat cells.
ELISA, Immuno Blotting, Immunoblot
Unconjugated
Purified
A recombinant polypeptide corresponding to amino acids of 71-170 human GRK6, expressed as a GST fusion protein
100μg
−20°C
Monoclonal
Human