Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GRK6, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232197

Catalog No. NBP232197


Add to cart

Description

Description

GRK6 Polyclonal antibody specifically detects GRK6 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

GRK6
1.0 mg/mL
Western Blot 1:2000 - 1:10000
NP_001004106.1
EC 2.7.11, EC 2.7.11.16, FLJ32135, G protein-coupled receptor kinase 6, GPRK6G protein-coupled receptor kinase GRK6
Rabbit
IgG
100 ul
Protein Kinase
Primary
2870
Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
GRK6
The immunogen this antibody was made to, maps to a region between residue 526 to 576 of human G Protein-Coupled Receptor Kinase 6 using the numbering given in entry NP_001004106.1 (GeneID 2870).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only