Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL1, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA120861
Description
The GRO gene was originally identified by subtractive hybridization studies between normal and tumorigenic Chinese hamster embryo fibroblasts. The hamster cDNA was cloned and used as a probe for cloning of the human GRO cDNA. The GROalpha gene initially cloned from T24 cells and the gene in melanoma cells encoding melanoma growth stimulating protein (MGSA) are identical.
PA1-20861 detects GRO-alpha in human, mouse, and rat samples.PA1-20861 has been successfully used in ELISA, and Western Blot procedures.PA1-20861 immunogen corresponds to Synthetic peptide corresponding to amino acids 35-107 of Human GRO alpha..Specifications
GRO alpha | |
Polyclonal | |
Unconjugated | |
P09341, P12850, P14095 | |
CXCL1 | |
Synthetic peptide corresponding to amino acids 35-107 of Human GRO alpha. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
ELISA, Western Blot | |
1mg/mL | |
CXCL1 | |
FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
2919 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction