Learn More
MilliporeSigma™ anti-Growth Associated Protein 43, Polyclonal, (Chemicon) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB9854/DEL
Catalog No. 50-174-291
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
The protein encoded by this gene has been termed a 'growth' or 'plasticity' protein because it is expressed at high levels in neuronal growth cones during development and axonal regeneration. This encoded protein is considered a crucial component of an effective regenerative response in the nervous system.
Western Blotting
Specifications
Specifications
Growth Associated Protein 43
Affinity Purified
Synthetic peptide
RUO
Primary
Murine
Unconjugated
Rabbit
100μg
Neuroscience
Polyclonal