Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRP78/HSPA5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 25 publications
Supplier: Novus Biologicals NBP106274SS
Description
GRP78/HSPA5 Polyclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat, C. elegans, Chicken, Drosophila, Sheep, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry - Free Floating.Specifications
GRP78/HSPA5 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
Rabbit | |
78 kDa | |
0.025 mL | |
Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
3309 | |
Human, Mouse, Rat, C. elegans, Chicken, Drosophila, Sheep | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating) | |
1.0 mg/mL | |
Western Blot 0.5 ug/ml, Simple Western 1:25, Flow Cytometry 1:150. Use reported in scientific literature (PMID 20208072), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50. Use reported in scientific literature (PMID 24089213), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24089213), Immunohistochemistry Free-Floating reported in scientific literature (PMID 26329458) | |
P11021 | |
HSPA5 | |
Synthetic peptide made to an internal portion of human GRP78 (within residues 250-300). [Swiss-Prot# P11021] | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, rat, sheep and chicken. Predicted to react with Zebrafish, S. cerevisiae, primate, bovine, and Drosophila based on 100% sequence homology. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction