Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GS15 Mouse, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

$232.00 - $427.00

Specifications

Antigen GS15
Isotype IgG1
Immunogen Rat GS15 aa. 1-85
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610960
View Documents Promotion Details
BD Biosciences
610960
50μg
Each for $232.00
Add to cart
 
BDB610961
View Documents Promotion Details
BD Biosciences
610961
150μg
Each for $427.00
Add to cart
 
Description

Description

Eukaryotic protein trafficking involves the packaging of target molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their components into an acceptor compartment. Components of both the vesicle and the synaptic plasma membrane interact to form a fusion complex which mediates specific docking and fusion of vesicles. This complex contains NSF (N-ethyl-maleimide-sensitive factor), SNAPs (soluble NSF attachment proteins), and receptor proteins (SNAREs) that include synaptobrevin, synaptotagmin, syntaxin, and SNAP-25 (synaptosome-associated protein of 25kDa). SNAP-25 and syntaxin are associated with the target plasma membrane (t-SNAREs), while synaptobrevin and synaptotagmin are vesicle-associated proteins (v-SNAREs). In Drosophila, protein transport from the ER to Golgi involves Bet1p, a v-SNARE. With 28% amino acid identity to Bet1p, GS15 (Golgi SNARE of 15kDa) is widely expressed in rat tissues. It is an integral membrane protein of the Golgi apparatus and functions as a SNARE. Thus, GS15 is thought to be a novel SNARE that participates in ER-Golgi protein transport through an undefined process.

Immunofluorescence, Western Blotting

Specifications

Specifications

GS15
Rat GS15 aa. 1-85
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Golgi SNARE of 15 kDa
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
19
250μg/mL
Store undiluted at -20°C.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit