Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GS27 Mouse, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611035

 View more versions of this product

Catalog No. BDB611035

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Eukaryotic protein trafficking involves the packaging of target molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their components into an acceptor compartment. Components of both the vesicle and the synaptic plasma membrane interact to form a fusion complex that mediates vesicle docking and fusion. This fusion complex contains NSF (N-ethyl-maleimide-sensitive factor), SNAPs (soluble NSF attachment proteins), and receptor proteins (SNAREs) that include synaptobrevin, synaptotagmin, syntaxin, and SNAP-25 (synaptosome-associated protein of 25kDa). Interactions between vesicle SNAREs and target membrane SNARES mediates the specificity of docking. Along one pathway of protein trafficking, the Golgi apparatus receives proteins from the ER. These proteins move from cis-Golgi to trans-Golgi, through a stack of cisternae, towards the trans-Golgi network. From here, they are delivered to their proper destination in the cell. GS27 (Golgi SNARE of 27kDa) (membrin) is a Golgi-associated SNARE. It functions in medial-to-trans-Golgi protein movement.

Immunofluorescence, Western Blotting

Specifications

Specifications

GS27
25
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Immunofluorescence, Western Blot
250μg/mL
Golgi SNARE of 27 kDa; Membrin
Affinity Purified
Human GS27 aa. 5-124
150μg
Cell Biology
Primary
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.