Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GS28 Mouse, Unlabeled, Clone: 1, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611185

 View more versions of this product

Catalog No. BDB611185


Add to cart

Description

Description

Most vesicular fusion events require the function of N-ethylmaleimide-sensitive factor (NSF) and the soluble NSF attachment proteins (SNAPs). Their function is dependent upon the SNAP receptor (SNARE) family of proteins. The specificity of vesicle docking and fusion is mediated by specific interactions between v-SNAREs on vesicles and t-SNAREs on target membranes. GS28 (Golgi SNARE with a size of 28kDa) is a SNARE that associates with the cis-Golgi and participates in trafficking between the ER and the Golgi and between Golgi
compartments. The majority of GS28, the first 230 and 250 aa, is thought to be anchored to the membrane via the C-terminal hydrophobic tail, which is formed by the remaining 20 aa. GS28 and syntaxin 5, another SNARE, exist as a protein complex in the Golgi and this complex is important for the function of both proteins in ER-Golgi transport. The GS28/syntaxin 5 complex can be dissociated by α-SNAP and NSF. In addition, GS28 is thought to interact with α-SNAP when the GS28/syntaxin 5 complex is dissociated. Thus, GS28 is a SNARE protein that mediates, in complex with syntaxin 5, transport within the Golgi and between Golgi and ER.

Host Species: Mouse
Clone: 1
Isotype: IgG2a
Species Reactivity: Mouse
Immunogen: Rat GS28 aa. 3-108
Formula Weight [Chemical]: 28kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

GS28
1
Unconjugated
Affinity Purified
Rat GS28 aa. 3-108
150μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.