Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GSAP, DyLight 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178376G

Catalog No. NBP178376G


Add to cart

Description

Description

GSAP Polyclonal antibody specifically detects GSAP in Human samples. It is validated for Western Blot.
Specifications

Specifications

GSAP
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
PION
A synthetic peptide made to an internal portion of the human GSAP protein (between residues 100-150) [UniProt A4D1B5]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human
Western Blot
Western Blot
A4D1B5
pigeon homolog (Drosophila)
Rabbit
IgG
0.1 ml
Alzheimers Research, Neurodegeneration, Neuroscience
Primary
54103
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only