Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GSK-3 alpha (p Thr202, p Tyr204), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP145784

Catalog No. NBP145784


Add to cart

Description

Description

GSK-3 alpha p Thr202, p Tyr204 Polyclonal antibody specifically detects GSK-3 alpha p Thr202, p Tyr204 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

GSK-3 alpha (p Thr202, p Tyr204)
0.5 mg/ml
Western Blot, Immunohistochemistry
NP_063937
DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha
Rabbit
IgG
0.1 mg
Cell Biology, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers
Primary
2931
Immunohistochemistry, Western Blot
Unconjugated
PBS (pH 7.2), 1% BSA, 50% Glycerol with 0.02% Sodium Azide
Affinity Purified
GSK3A
Synthetic phospho-peptide derived from human GSK-3a.
Affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only